Learn More
Sorbic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Sorbic acid ethyl ester |
| CAS | 2396-84-1 |
| Molecular Formula | C8 H12 O2 |
| Quantity | 10 g |
| Synonym | Ethyl Sorbate, (2E,4E)-2,4-Hexadienoic acid ethyl ester, 2,4-Hexadienoic acid','ethyl ester','(E,E)-, Sorbic acid','ethyl ester (6CI,7CI,8CI), (E,E)-Ethyl 2,4-hexadienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, Ethyl 2,4-hexadienoate, Ethyl sorbate, NSC 8874 |
| InChI Formula | InChI=1S/C8H12O2/c1-3-5-6-7-8(9)10-4-2/h3,5-7H,4H2,1-2H3/b5-3+,7-6+ |
| SMILES | CCOC(=O)\C=C\C=C\C |
| IUPAC Name | ethyl (2E,4E)-hexa-2,4-dienoate |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.0837 |