missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid Ethyl Ester, TRC

Product Code. 30413830
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413830 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413830 Supplier Toronto Research Chemicals Supplier No. TRCS676895100G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sorbic acid ethyl ester
CAS 2396-84-1
Molecular Formula C8 H12 O2
Quantity 100 g
Synonym Ethyl Sorbate, (2E,4E)-2,4-Hexadienoic acid ethyl ester, 2,4-Hexadienoic acid','ethyl ester','(E,E)-, Sorbic acid','ethyl ester (6CI,7CI,8CI), (E,E)-Ethyl 2,4-hexadienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, Ethyl 2,4-hexadienoate, Ethyl sorbate, NSC 8874
InChI Formula InChI=1S/C8H12O2/c1-3-5-6-7-8(9)10-4-2/h3,5-7H,4H2,1-2H3/b5-3+,7-6+
SMILES CCOC(=O)\C=C\C=C\C
IUPAC Name ethyl (2E,4E)-hexa-2,4-dienoate
Molecular Weight (g/mol) 140.18
Formula Weight 140.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.