CAS RN 23358-95-4
CAS RN 23358-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Bis(4-hydroxybutyl) Terephthalate, TRC
CAS: 23358-95-4 Molecular Formula: C16 H22 O6 Molecular Weight (g/mol): 310.34 Synonym: 1,4-Benzenedicarboxylic acid 1,4-bis(4-hydroxybutyl) ester,1,4-Benzenedicarboxylic acid, bis(4-hydroxybutyl) ester (9CI),Terephthalic acid, bis(4-hydroxybutyl) ester (6CI,8CI),1,4-Butanediol, terephthalate (2:1) (8CI),1,4-Butanediyl terephthalate,Bis(4-hydroxybutyl) terephthalate IUPAC Name: bis(4-hydroxybutyl) benzene-1,4-dicarboxylate SMILES: OCCCCOC(=O)c1ccc(cc1)C(=O)OCCCCO
2
Bis(4-hydroxybutyl) Terephthalate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.