missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxybutyl) Terephthalate, TRC

Product Code. 30338798
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338798 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338798 Supplier Toronto Research Chemicals Supplier No. TRCB44381550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(4-hydroxybutyl) Terephthalate
CAS 23358-95-4
Purity Grade Notes HPLC
Molecular Formula C16 H22 O6
Quantity 50 mg
Synonym 1,4-Benzenedicarboxylic acid 1,4-bis(4-hydroxybutyl) ester, 1,4-Benzenedicarboxylic acid, bis(4-hydroxybutyl) ester (9CI), Terephthalic acid, bis(4-hydroxybutyl) ester (6CI,8CI), 1,4-Butanediol, terephthalate (2:1) (8CI), 1,4-Butanediyl terephthalate, Bis(4-hydroxybutyl) terephthalate
InChI Formula InChI=1S/C16H22O6/c17-9-1-3-11-21-15(19)13-5-7-14(8-6-13)16(20)22-12-4-2-10-18/h5-8,17-18H,1-4,9-12H2
SMILES OCCCCOC(=O)c1ccc(cc1)C(=O)OCCCCO
IUPAC Name bis(4-hydroxybutyl) benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 310.34
Formula Weight 310.1416
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.