Learn More
Bis(4-hydroxybutyl) Terephthalate, TRC
Product Code. 17959194
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959194 10 mg 10mg
17917654 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17959194 Supplier Toronto Research Chemicals Supplier No. TRCB44381510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23358-95-4
Molecular Formula C16 H22 O6
Molecular Weight (g/mol) 310.34
Synonym 1,4-Benzenedicarboxylic acid 1,4-bis(4-hydroxybutyl) ester, 1,4-Benzenedicarboxylic acid, bis(4-hydroxybutyl) ester (9CI), Terephthalic acid, bis(4-hydroxybutyl) ester (6CI,8CI), 1,4-Butanediol, terephthalate (2:1) (8CI), 1,4-Butanediyl terephthalate, Bis(4-hydroxybutyl) terephthalate
IUPAC Name bis(4-hydroxybutyl) benzene-1,4-dicarboxylate
SMILES OCCCCOC(=O)c1ccc(cc1)C(=O)OCCCCO
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H22O6/c17-9-1-3-11-21-15(19)13-5-7-14(8-6-13)16(20)22-12-4-2-10-18/h5-8,17-18H,1-4,9-12H2
Formula Weight 310.1416
Percent Purity >95%
Chemical Name or Material Bis(4-hydroxybutyl) Terephthalate
Grade HPLC