accessibility menu, dialog, popup

UserName

CH3SOOHNONNNHOFCl

CAS RN 231277-92-2

IUPAC Name: N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-(5-{[(2-methanesulfonylethyl)amino]methyl}furan-2-yl)quinazolin-4-amine
Synonyms: lapatinib tykerb lapatinib ditosylate tycerb C29H26ClFN4O4S lapatinib tosilate hydrate FMM
Molecular Weight (g/mol): 581.06
Molecular Formula: C29H26ClFN4O4S
InChi Key: BCFGMOOMADDAQU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CCNCC1=CC=C(O1)C1=CC=C2N=CN=C(NC3=CC=C(OCC4=CC=CC(F)=C4)C(Cl)=C3)C2=C1
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results