CAS RN 2306-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Sinensetin, TRC
CAS: 2306-27-6 Molecular Formula: C20 H20 O7 Molecular Weight (g/mol): 372.37 Synonym: 2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxy-4H-1-benzopyran-4-one,3',4',5,6,7-Pentamethoxyflavone,5,6,7,3',4'-Pentamethoxyflavone,Pedalitin Permethyl Ether; IUPAC Name: 2-(3,4-dimethoxyphenyl)-5,6,7-trimethoxychromen-4-one SMILES: COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)cc3O2
2
Sinensetin, MedChemExpress
MedChemExpress Sinensetin is a methylated flavone found in certain citrus fruits.