missing translation for 'onlineSavingsMsg'
Learn More

Sinensetin, MedChemExpress

Product Code. 16488938
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488938 10 mg 10mg
16441378 5 mg 5mg
16433856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488938 Supplier Medchem Express Supplier No. HYN0297
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Sinensetin is a methylated flavone found in certain citrus fruits.

Sinensetin is a methylated flavone found in certain citrus fruits.

Specifications

Chemical Name or Material Sinensetin
CAS 2306-27-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H20O7
Quantity 10 mg
Synonym Pedalitin permethyl ether
Solubility Information DMSO : 100 mg/mL (268.55 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=CC(OC)=C(OC)C(OC)=C13
Molecular Weight (g/mol) 372.37
Formula Weight 372.37
Percent Purity 99.34%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.