missing translation for 'onlineSavingsMsg'
Learn More

Sinensetin, TRC

Product Code. 30008917
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008917 10 mg 10mg
30010377 100 mg 100mg
30005304 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008917 Supplier Toronto Research Chemicals Supplier No. TRCS48730010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2306-27-6
Molecular Formula C20 H20 O7
Molecular Weight (g/mol) 372.37
Synonym 2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxy-4H-1-benzopyran-4-one, 3',4',5,6,7-Pentamethoxyflavone, 5,6,7,3',4'-Pentamethoxyflavone, Pedalitin Permethyl Ether;
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,6,7-trimethoxychromen-4-one
SMILES COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)cc3O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H20O7/c1-22-13-7-6-11(8-15(13)23-2)14-9-12(21)18-16(27-14)10-17(24-3)19(25-4)20(18)26-5/h6-10H,1-5H3
Formula Weight 372.1209
Percent Purity >95%
Chemical Name or Material Sinensetin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.