CAS RN 22871-77-8
IUPAC Name:
5,5-dimethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborinane
Synonyms:
5,5-dimethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborinane
Molecular Weight (g/mol):
240.11
Molecular Formula:
C15H17BO2
InChi Key:
VWRKQVBSFBSIOI-UHFFFAOYSA-N
SMILES:
CC1(C)COB(OC1)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
1-Naphthaleneboronic acid neopentyl glycol ester, 98%, Thermo Scientific™
CAS: 22871-77-8 Molecular Formula: C15H17BO2 Molecular Weight (g/mol): 240.109 MDL Number: MFCD01318167 InChI Key: VWRKQVBSFBSIOI-UHFFFAOYSA-N Synonym: 1-naphthaleneboronic acid neopentyl glycol ester,5,5-dimethyl-2-naphthalen-1-yl-1,3,2-dioxaborinane,1-naphthaleneboronic acid neopentyl glycol cyclic ester,2-1-naphthyl-5,5-dimethyl-1,3,2-dioxaborinane,5,5-dimethyl-2-alpha-naphthyl-1,3,2-dioxaborinane PubChem CID: 22945154 IUPAC Name: 5,5-dimethyl-2-naphthalen-1-yl-1,3,2-dioxaborinane SMILES: B1(OCC(CO1)(C)C)C2=CC=CC3=CC=CC=C23