missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic acid neopentyl glycol ester, 98%, Thermo Scientific™

Product Code. 11368556
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368556 5 g 5g
11358556 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368556 Supplier Thermo Scientific Alfa Aesar Supplier No. L17612.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22871-77-8
Molecular Formula C15H17BO2
Molecular Weight (g/mol) 240.109
MDL Number MFCD01318167
InChI Key VWRKQVBSFBSIOI-UHFFFAOYSA-N
Synonym 1-naphthaleneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-naphthalen-1-yl-1,3,2-dioxaborinane, 1-naphthaleneboronic acid neopentyl glycol cyclic ester, 2-1-naphthyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-alpha-naphthyl-1,3,2-dioxaborinane
PubChem CID 22945154
IUPAC Name 5,5-dimethyl-2-naphthalen-1-yl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=CC3=CC=CC=C23

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
Beilstein 3053756
Formula Weight 240.11
Percent Purity 98%
Chemical Name or Material 1-Naphthaleneboronic acid neopentyl glycol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.