accessibility menu, dialog, popup

UserName

H3CSCH3

CAS RN 22438-39-7

IUPAC Name: 1-(methylsulfanyl)decane
Synonyms: 1-(methylsulfanyl)decane
Molecular Weight (g/mol): 188.37
Molecular Formula: C11H24S
InChi Key: HKGUUZAACYBIID-UHFFFAOYSA-N
SMILES: CCCCCCCCCCSC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)