missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Decyl methyl sulfide, 97%

Product Code. 11397455
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397455 10 g 10g
11307465 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397455 Supplier Alfa Aesar™ Supplier No. L05473.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22438-39-7
Molecular Formula C11H24S
Molecular Weight (g/mol) 188.37
MDL Number MFCD00026541
InChI Key HKGUUZAACYBIID-UHFFFAOYSA-N
Synonym decyl methyl sulfide, n-decyl methyl sulfide, decyl methyl sulphide, 1-methylsulfanyl decane, 2-thiadodecane, decane, 1-methylthio, decylmethyl sulfide, n-decyl methyl sulphide, acmc-209fwh
PubChem CID 89712
IUPAC Name 1-methylsulfanyldecane
SMILES CCCCCCCCCCSC

Specifications

Density 0.845
Boiling Point 126°C to 127°C (14 mmHg)
Flash Point 101°C (214°F)
Linear Formula CH3(CH2)9SCH3
Refractive Index 1.4605
Quantity 10 g
Beilstein 1736530
Formula Weight 188.37
Percent Purity 97%
Chemical Name or Material n-Decyl methyl sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.