accessibility menu, dialog, popup

UserName

HOOCl

CAS RN 21640-48-2

IUPAC Name: 3-(3-chlorophenyl)propanoic acid
Synonyms: 3-(3-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: CLTDVBQNUHHYCA-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results