missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Chlorophenyl)propionic acid, 96%

Product Code. 15496077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496077 25 g 25g
15476077 1 g 1g
15486077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496077 Supplier Thermo Scientific Alfa Aesar Supplier No. H33266.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21640-48-2 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21640-48-2
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD01310801
InChI Key CLTDVBQNUHHYCA-UHFFFAOYSA-N
Synonym 3-3-chlorophenyl propanoic acid, 3-3-chlorophenyl propionic acid, benzenepropanoic acid, 3-chloro, 3-chlorobenzenepropanoic acid, 3-3-chloro-phenyl-propionic acid, 3-chlorophenyl-3-propanoic acid, 3-chlorohydrocinnamic acid, acmc-1ch2q, benzenepropanoic acid,3-chloro, 3-3-chlorophenyl propanoicacid
PubChem CID 177164
IUPAC Name 3-(3-chlorophenyl)propanoic acid
SMILES OC(=O)CCC1=CC=CC(Cl)=C1

Specifications

Quantity 25 g
Formula Weight 184.62
Percent Purity 96%
Chemical Name or Material 3-(3-Chlorophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.