accessibility menu, dialog, popup

UserName

CAS RN 21436-98-6

HClCH3H3CH2N

CAS RN 21436-98-6

IUPAC Name: hydrogen 2,6-dimethylaniline chloride
Synonyms: 2,6-xylidine, hydrochloride
Molecular Weight (g/mol): 157.64
Molecular Formula: C8H12ClN
InChi Key: QNWMFJDRMZWZNN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results