Learn More
2,6-Dimethylaniline Hydrochloride, TRC
Product Code. 30009721
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009721 25 mg 25mg
30008087 1 g 1g
30007265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009721 Supplier Toronto Research Chemicals Supplier No. TRCD46220025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21436-98-6
Molecular Formula C8 H11 N . Cl H
Molecular Weight (g/mol) 157.64
Synonym Benzenamine, 2,6-dimethyl-, hydrochloride (1:1) (ACI), 2,6-Xylidine, hydrochloride (8CI), Benzenamine, 2,6-dimethyl-, hydrochloride (9CI), 2,6-Dimethylaniline hydrochloride, 2,6-Dimethylbenzenamine hydrochloride
IUPAC Name 2,6-dimethylaniline;hydrochloride
SMILES Cl.Cc1cccc(C)c1N
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C8H11N.ClH/c1-6-4-3-5-7(2)8(6)9;/h3-5H,9H2,1-2H3;1H
Formula Weight 157.0658
Percent Purity >95%
Chemical Name or Material 2,6-dimethylaniline hydrochloride
Grade HPLC