missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline Hydrochloride, Mikromol™

Product Code. 17941196
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941196 Supplier Mikromol Supplier No. MM0102.150025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,6-Dimethylaniline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-dimethylaniline hydrochloride
CAS 21436-98-6
Analyte or Component Names 2,6-dimethylaniline hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H11 N . Cl H
Certifications/Compliance ISO 17025
Synonym 2,6-Dimethylaniline Hydrochloride, Lidocaine Hydrochloride Imp. A (EP), Lidocaine Imp. A (EP), Ropivacaine USP Related Compound A, Ropivacaine USP RC A, Ropivacaine Related Compound A as Hydrochloride, Lidocaine Hydrochloride Impurity A as Hydrochloride, Lidocaine Impurity A as Hydrochloride
InChI Formula InChI=1S/C8H11N.ClH/c1-6-4-3-5-7(2)8(6)9;/h3-5H,9H2,1-2H3;1H
SMILES Cl.Cc1cccc(C)c1N
IUPAC Name 2,6-dimethylaniline;hydrochloride
Molecular Weight (g/mol) 157.64
Quantity 25 mg
Formula Weight 157.0658 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.