accessibility menu, dialog, popup

UserName

CAS RN 209984-56-5

IUPAC Name: 2-[2-(3,5-difluorophenyl)acetamido]-N-{8-methyl-9-oxo-8-azatricyclo[9.4.0.0²,⁷]pentadeca-1(15),2,4,6,11,13-hexaen-10-yl}propanamide
Synonyms: 2-[2-(3,5-difluorophenyl)acetamido]-N-{8-methyl-9-oxo-8-azatricyclo[9.4.0.0²,⁷]pentadeca-1(15),2,4,6,11,13-hexaen-10-yl}propanamide
Molecular Weight (g/mol): 463.49
Molecular Formula: C26H23F2N3O3
InChi Key: QSHGISMANBKLQL-UHFFFAOYNA-N
SMILES: CC(NC(=O)CC1=CC(F)=CC(F)=C1)C(=O)NC1C2=CC=CC=C2C2=CC=CC=C2N(C)C1=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (5)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)