CAS RN 20766-85-2
IUPAC Name:
6-(prop-2-enamido)hexanoic acid
Synonyms:
6-(prop-2-enamido)hexanoic acid
Molecular Weight (g/mol):
185.22
Molecular Formula:
C9H15NO3
InChi Key:
SAQWCPXBLNGTCC-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCNC(=O)C=C
1
–
3
of
3
results
1
6-Acrylamidohexanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Acrylamidohexanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Acrylamidohexanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.