missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Acrylamidohexanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Acrylamidohexanoic Acid |
| CAS | 20766-85-2 |
| Molecular Formula | C9 H15 N O3 |
| Quantity | 100 mg |
| Synonym | 6-[(1-Oxo-2-propen-1-yl)amino]hexanoic Acid, 6-Acrylamidocaproic Acid, 6-Acryloylaminocaproic Acid, 6-Acryloylaminohexanoic Acid, N-Acryloyl-6-aminocaproic Acid, N-Acryloyl-6-aminohexanoic Acid, N-Acryloyl-ε-amino-n-caproic Acid, N-Acryloyl-ω-aminohexanoic Acid, NSC 288649, ε-(Acryloylamino)caproic Acid |
| InChI Formula | InChI=1S/C9H15NO3/c1-2-8(11)10-7-5-3-4-6-9(12)13/h2H,1,3-7H2,(H,10,11)(H,12,13) |
| SMILES | OC(=O)CCCCCNC(=O)C=C |
| IUPAC Name | 6-(prop-2-enoylamino)hexanoic acid |
| Molecular Weight (g/mol) | 185.22 |
| Formula Weight | 185.105 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?