CAS RN 20261-38-5
CAS RN 20261-38-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ginkgolic Acid, TRC
CAS: 20261-38-5 Molecular Formula: C20 H32 O3 Molecular Weight (g/mol): 320.47 Synonym: Benzoic acid, 2-hydroxy-6-tridecyl-,Salicylic acid, 6-tridecyl- (8CI),2-Hydroxy-6-tridecylbenzoic acid,6-Tridecylsalicylic acid,6-n-Tridecylsalicylic acid,Ginkgoneolic acid IUPAC Name: 2-hydroxy-6-tridecylbenzoic acid SMILES: CCCCCCCCCCCCCc1cccc(O)c1C(=O)O
2
Ginkgolic Acid (C13:0), MedChemExpress
MedChemExpress Ginkgolic Acid (C13:0) is a natural anticariogenic agent in that it exhibits antimicrobial activity against S.