missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolic Acid, TRC

Product Code. 17991165
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991165 10 mg 10mg
17964155 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991165 Supplier Toronto Research Chemicals Supplier No. TRCG38740010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20261-38-5
Molecular Formula C20 H32 O3
Molecular Weight (g/mol) 320.47
Synonym Benzoic acid, 2-hydroxy-6-tridecyl-, Salicylic acid, 6-tridecyl- (8CI), 2-Hydroxy-6-tridecylbenzoic acid, 6-Tridecylsalicylic acid, 6-n-Tridecylsalicylic acid, Ginkgoneolic acid
IUPAC Name 2-hydroxy-6-tridecylbenzoic acid
SMILES CCCCCCCCCCCCCc1cccc(O)c1C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H32O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-15-13-16-18(21)19(17)20(22)23/h13,15-16,21H,2-12,14H2,1H3,(H,22,23)
Formula Weight 320.2351
Chemical Name or Material Ginkgolic Acid C13:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.