missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolic Acid (C13:0), MedChemExpress

Product Code. 16428858
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428858 Supplier Medchem Express Supplier No. HYN0078
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ginkgolic Acid (C13:0) is a natural anticariogenic agent in that it exhibits antimicrobial activity against S.

Ginkgolic Acid (C13:0) is a natural anticariogenic agent in that it exhibits antimicrobial activity against S.

Specifications

Chemical Name or Material Ginkgolic Acid (C13:0)
CAS 20261-38-5
Color White
Purity Grade Notes Research
Molecular Formula C20H32O3
Quantity 10 mg
Synonym Ginkgolic acid (13:0) Ginkgoneolic Acid 6-Tridecylsalicylic acid
Solubility Information DMSO : 100 mg/mL (312.04 mM; Need ultrasonic)
SMILES O=C(O)C1=C(CCCCCCCCCCCCC)C=CC=C1O
Molecular Weight (g/mol) 320.47
Formula Weight 320.47
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.