CAS RN 18750-05-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzyl 2-Ethylhexyl Phthalate, TRC
CAS: 18750-05-5 Molecular Formula: C23 H28 O4 Molecular Weight (g/mol): 368.47 Synonym: 1,2-Benzenedicarboxylic acid, 2-ethylhexyl phenylmethyl ester (9CI),Phthalic acid, benzyl 2-ethylhexyl ester (6CI,7CI,8CI),2-Ethylhexyl benzyl phthalate,Benzyl 2-ethylhexyl phthalate,Benzyl octyl phthalate,OBzP,Octyl benzyl phthalate,Placizer B 8 IUPAC Name: 1-O-benzyl 2-O-(2-ethylhexyl) benzene-1,2-dicarboxylate SMILES: CCCCC(CC)COC(=O)c1ccccc1C(=O)OCc2ccccc2