missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Ethylhexyl Phthalate, TRC

Product Code. 30135860
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135860 5 g 5g
30132566 2.5 g 2.5g
30006130 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135860 Supplier Toronto Research Chemicals Supplier No. TRCB4108535G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18750-05-5
Molecular Formula C23 H28 O4
Molecular Weight (g/mol) 368.47
Synonym 1,2-Benzenedicarboxylic acid, 2-ethylhexyl phenylmethyl ester (9CI), Phthalic acid, benzyl 2-ethylhexyl ester (6CI,7CI,8CI), 2-Ethylhexyl benzyl phthalate, Benzyl 2-ethylhexyl phthalate, Benzyl octyl phthalate, OBzP, Octyl benzyl phthalate, Placizer B 8
IUPAC Name 1-O-benzyl 2-O-(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)c1ccccc1C(=O)OCc2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C23H28O4/c1-3-5-11-18(4-2)16-26-22(24)20-14-9-10-15-21(20)23(25)27-17-19-12-7-6-8-13-19/h6-10,12-15,18H,3-5,11,16-17H2,1-2H3
Formula Weight 368.1988
Chemical Name or Material Phthalic acid, benzyl-2-ethylhexyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.