CAS RN 18328-90-0
CAS RN 18328-90-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Ethyl-2-methyl-2-propen-1-amine, TRC
CAS: 18328-90-0 Molecular Formula: C6H13N Molecular Weight (g/mol): 99.17 Synonym: N-Ethyl-2-methyl-allylamine,N-2-Methylallyl-N-ethylamine,N-Ethyl(2-methyl-2-propenyl)amine,N-Ethyl-2-methylallylamine,N-Ethylmethallylamine SMILES: CCNCC(=C)C