Learn More
N-Ethyl-2-methyl-2-propen-1-amine, TRC
Product Code. 18042250
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042250 250 mg 250mg
18030450 50 mg 50mg
18001950 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18042250 Supplier Toronto Research Chemicals Supplier No. TRCE925078250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18328-90-0
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.17
Synonym N-Ethyl-2-methyl-allylamine, N-2-Methylallyl-N-ethylamine, N-Ethyl(2-methyl-2-propenyl)amine, N-Ethyl-2-methylallylamine, N-Ethylmethallylamine
SMILES CCNCC(=C)C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H13N/c1-4-7-5-6(2)3/h7H,2,4-5H2,1,3H3
Formula Weight 99.1048
Chemical Name or Material N-Ethyl-2-methyl-2-propen-1-amine