accessibility menu, dialog, popup

UserName

CH3OH3COCH3

CAS RN 18229-78-2

IUPAC Name: 1,1-diethoxyhex-2-yne
Synonyms: 1,1-diethoxyhex-2-yne
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: VUNHOKCOJYJVBT-UHFFFAOYSA-N
SMILES: CCCC#CC(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results