missing translation for 'onlineSavingsMsg'
Learn More

2-Hexynal diethyl acetal, 98%, Thermo Scientific™

Product Code. 11382366
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382366 25 g 25g
11372366 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382366 Supplier Thermo Scientific Alfa Aesar Supplier No. L06098.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18229-78-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00041643
InChI Key VUNHOKCOJYJVBT-UHFFFAOYSA-N
Synonym 2-hexynal diethyl acetal, 2-hexyne, 1,1-diethoxy, 1,1-diethoxy-2-hexyne, acmc-1c7c9, 2-hexyn-1-al diethyl acetal, 2-hexynyl aldehyde diethyl acetal
PubChem CID 87518
IUPAC Name 1,1-diethoxyhex-2-yne
SMILES CCCC#CC(OCC)OCC

Specifications

Density 0.885
Boiling Point 79°C to 80°C (8 mmHg)
Refractive Index 1.431
Quantity 25 g
Beilstein 1703624
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material 2-Hexynal diethyl acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.