CAS RN 181289-23-6
CAS RN 181289-23-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(S)-Pregabalin Lactam, TRC
CAS: 181289-23-6 Molecular Formula: C8 H15 N O Molecular Weight (g/mol): 141.21 Synonym: (4S)-4-(2-methylpropyl)-2-Pyrrolidinone,2-Pyrrolidinone, 4-(2-methylpropyl)-, (S)-,(S)-4-(Isobutyl)pyrrolidin-2-one,(S)-Pregabalin lactam,(4S)-4-(2-methylpropyl)-2-Pyrrolidinone,2-Pyrrolidinone, 4-(2-methylpropyl)-, (S)-,(S)-4-(Isobutyl)pyrrolidin-2-one,Pregabalin EP impurity A IUPAC Name: (4S)-4-(2-methylpropyl)pyrrolidin-2-one SMILES: CC(C)C[C@@H]1CNC(=O)C1
2
(4S)-4-(2-Methylpropyl)pyrrolidin-2-one ((S)-Pregabalin Lactam), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.