Learn More
(S)-Pregabalin Lactam, TRC
Product Code. 17990415
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990415 50 mg 50mg
17921885 250 mg 250mg
17911465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17990415 Supplier Toronto Research Chemicals Supplier No. TRCP70482050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 181289-23-6
Molecular Formula C8 H15 N O
Molecular Weight (g/mol) 141.21
Synonym (4S)-4-(2-methylpropyl)-2-Pyrrolidinone, 2-Pyrrolidinone, 4-(2-methylpropyl)-, (S)-, (S)-4-(Isobutyl)pyrrolidin-2-one, (S)-Pregabalin lactam, (4S)-4-(2-methylpropyl)-2-Pyrrolidinone, 2-Pyrrolidinone, 4-(2-methylpropyl)-, (S)-, (S)-4-(Isobutyl)pyrrolidin-2-one, Pregabalin EP impurity A
IUPAC Name (4S)-4-(2-methylpropyl)pyrrolidin-2-one
SMILES CC(C)C[C@@H]1CNC(=O)C1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8H15NO/c1-6(2)3-7-4-8(10)9-5-7/h6-7H,3-5H2,1-2H3,(H,9,10)/t7-/m0/s1
Formula Weight 141.1154
Percent Purity >95%
Chemical Name or Material (S)-Pregabalin Lactam
Grade HPLC