CAS RN 175205-51-3
IUPAC Name:
1-[3,5-bis(trifluoromethyl)phenyl]-2,5-dimethyl-1H-pyrrole
Synonyms:
1-[3,5-bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Molecular Weight (g/mol):
307.24
Molecular Formula:
C14H11F6N
InChi Key:
KUDHKDYSHKOCTC-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C)N1C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole, 97%, Alfa Aesar™
CAS: 175205-51-3 Molecular Formula: C14H11F6N Molecular Weight (g/mol): 307.24 MDL Number: MFCD00084902 InChI Key: KUDHKDYSHKOCTC-UHFFFAOYSA-N Synonym: 1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl-1h-pyrrole,1-3,5-bis trifluoromethyl phenyl-2,5-dimethylpyrrole,1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl-pyrrole,acmc-1bzzk,1-3,5-di trifluoromethyl phenyl-2,5-dimethyl-1h-pyrrole,1h-pyrrole,1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl PubChem CID: 1800795 SMILES: CC1=CC=C(C)N1C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F