missing translation for 'onlineSavingsMsg'
Learn More

1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole, 97%, Alfa Aesar™

Product Code. 11426097
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426097 25 g 25g
11406097 1 g 1g
11416097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426097 Supplier Thermo Scientific Alfa Aesar Supplier No. B25087.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-51-3
Molecular Formula C14H11F6N
Molecular Weight (g/mol) 307.24
MDL Number MFCD00084902
InChI Key KUDHKDYSHKOCTC-UHFFFAOYSA-N
Synonym 1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl-1h-pyrrole, 1-3,5-bis trifluoromethyl phenyl-2,5-dimethylpyrrole, 1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl-pyrrole, acmc-1bzzk, 1-3,5-di trifluoromethyl phenyl-2,5-dimethyl-1h-pyrrole, 1h-pyrrole,1-3,5-bis trifluoromethyl phenyl-2,5-dimethyl
PubChem CID 1800795
SMILES CC1=CC=C(C)N1C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 78°C to 80°C (0.1 mmHg)
Refractive Index 1.4675
Quantity 25 g
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Formula Weight 283.22
Percent Purity 97%
Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.