accessibility menu, dialog, popup

UserName

H3C

CAS RN 1705-85-7

IUPAC Name: 6-methylchrysene
Synonyms: 6-methylchrysene
Molecular Weight (g/mol): 242.32
Molecular Formula: C19H14
InChi Key: ASVDRLYVNFOSCI-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=CC3=CC=CC=C23)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results