missing translation for 'onlineSavingsMsg'
Learn More

6-Methylchrysene 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30324001
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324001 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324001 Supplier Dr. Ehrenstorfer Supplier No. DREYS09010018DI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Chemical Name or Material 6-Methylchrysene
CAS 1705-85-7
Quantity 5 x 1 mL
Analyte or Component Names 6-Methylchrysene
Shipping Condition Room Temperature
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Chrysene, 6-methyl-, 6-Methylchrysene
InChI Formula InChI=1S/C19H14/c1-13-12-19-16-8-3-2-6-14(16)10-11-18(19)17-9-5-4-7-15(13)17/h2-12H,1H3
Solution Type Single Solution
IUPAC Name 6-methylchrysene
Solvent or Matrix Dichloromethane
Industry Type Environmental Testing
Concentration or Composition (by Analyte or Components) 6-Methylchrysene 1000 μg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.