Learn More
6-Methyl Chrysene, TRC
Product Code. 30472719
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472719 100 mg 100mg
30467355 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30472719 Supplier Toronto Research Chemicals Supplier No. TRCM265140100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1705-85-7
Molecular Formula C19 H14
Molecular Weight (g/mol) 242.31
Synonym Chrysene, 6-methyl-, 6-Methylchrysene
IUPAC Name 6-methylchrysene
SMILES Cc1cc2c3ccccc3ccc2c4ccccc14
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C19H14/c1-13-12-19-16-8-3-2-6-14(16)10-11-18(19)17-9-5-4-7-15(13)17/h2-12H,1H3
Formula Weight 242.1096
Percent Purity >95%
Chemical Name or Material 6-Methylchrysene
Grade HPLC