CAS RN 14885-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ipronidazole, TRC
CAS: 14885-29-1 Molecular Formula: C7 H11 N3 O2 Molecular Weight (g/mol): 169.18 Synonym: Imidazole, 2-isopropyl-1-methyl-5-nitro- (8CI),1-Methyl-2-(1-methylethyl)-5-nitro-1H-imidazole,1-Methyl-2-isopropyl-5-nitroimidazole,2-Isopropyl-1-methyl-5-nitroimidazole,Ipronidazole,Ipropran,NSC 109212,Ro 07-1554 IUPAC Name: 1-methyl-5-nitro-2-propan-2-ylimidazole SMILES: CC(C)c1ncc([N+](=O)[O-])n1C
2
Ipronidazole (1000 ug/mL in Methanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.