missing translation for 'onlineSavingsMsg'
Learn More

Ipronidazole (1000 ug/mL in Methanol), TRC

Product Code. 30389004
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389004 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389004 Supplier Toronto Research Chemicals Supplier No. TRCKIT04681X1ML
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ipronidazole
CAS 14885-29-1
Molecular Formula C7 H11 N3 O2
Quantity 1x1 mL
Synonym Imidazole, 2-isopropyl-1-methyl-5-nitro- (8CI), 1-Methyl-2-(1-methylethyl)-5-nitro-1H-imidazole, 1-Methyl-2-isopropyl-5-nitroimidazole, 2-Isopropyl-1-methyl-5-nitroimidazole, Ipronidazole, Ipropran, NSC 109212, Ro 07-1554
InChI Formula InChI=1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3
SMILES CC(C)c1ncc([N+](=O)[O-])n1C
IUPAC Name 1-methyl-5-nitro-2-propan-2-ylimidazole
Molecular Weight (g/mol) 169.18
Formula Weight 169.0851
Concentration or Composition (by Analyte or Components) 1000 μg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.