missing translation for 'onlineSavingsMsg'
Learn More

Ipronidazole, TRC

Product Code. 17990345
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990345 100 mg 100mg
17914005 50 mg 50mg
17921615 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990345 Supplier Toronto Research Chemicals Supplier No. TRCI747000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14885-29-1
Molecular Formula C7 H11 N3 O2
Molecular Weight (g/mol) 169.18
Synonym Imidazole, 2-isopropyl-1-methyl-5-nitro- (8CI), 1-Methyl-2-(1-methylethyl)-5-nitro-1H-imidazole, 1-Methyl-2-isopropyl-5-nitroimidazole, 2-Isopropyl-1-methyl-5-nitroimidazole, Ipronidazole, Ipropran, NSC 109212, Ro 07-1554
IUPAC Name 1-methyl-5-nitro-2-propan-2-ylimidazole
SMILES CC(C)c1ncc([N+](=O)[O-])n1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3
Formula Weight 169.0851
Percent Purity >95%
Chemical Name or Material Ipronidazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.