accessibility menu, dialog, popup

UserName

ONOHNNHOSOCl

CAS RN 146032-79-3

IUPAC Name: 6-chloro-N'-(3-{[(furan-2-yl)methyl]sulfanyl}propanoyl)-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-9-carbohydrazide
Synonyms: 6-chloro-N'-{3-[(furan-2-ylmethyl)sulfanyl]propanoyl}-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-9-carbohydrazide
Molecular Weight (g/mol): 457.93
Molecular Formula: C22H20ClN3O4S
InChi Key: CQBVTZDISUKDSX-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(OC3=CC=CC=C3CN2C(=O)NNC(=O)CCSCC2=CC=CO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results