accessibility menu, dialog, popup

UserName

CAS RN 146-56-5

HHClClOHNNNSFFF

CAS RN 146-56-5

IUPAC Name: dihydrogen 2-(4-{3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl}piperazin-1-yl)ethan-1-ol dichloride
Synonyms: dihydrogen fluphenazine dichloride
Molecular Weight (g/mol): 510.44
Molecular Formula: C22H28Cl2F3N3OS
InChi Key: MBHNWCYEGXQEIT-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].OCCN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1
Industry Type 
  • (1)
  • (1)
  • (4)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results