missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluphenazine dihydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Fluphenazine dihydrochloride is a phenothiazine-class D1DR and D2DR inhibitor
Specifications
Specifications
| Chemical Name or Material | Fluphenazine dihydrochloride |
| Physical Form | Powder |
| CAS | 146-56-5 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H28Cl2F3N3OS |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | H2O : 100 mg/mL (195.91 mM; Need ultrasonic) ∣DMSO : ≥ 38 mg/mL (74.45 mM) |
| SMILES | OCCN1CCN(CCCN2C3=C(C=CC=C3)SC4=CC=C(C(F)(F)F)C=C24)CC1.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction