missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine.2HCl, Lipomed

Product Code. 30459037
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30459037

Brand: Lipomed LPMFPH1233HC100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Fluphenazine.2HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluphenazine dihydrochloride
CAS 146-56-5
Analyte or Component Names Fluphenazine dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H26 F3 N3 O S . 2 Cl H
Synonym Fluphenazine Dihydrochloride, 4-[3-[2-(Trifluoromethyl)-10 H-phenothiazin-10-yl]propyl]-1-piperazineethanol hydrochloride (1:2), 4-[3-[2-(Trifluoromethyl)-10 H-phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, 1-(2-Hydroxyethyl)-4-[3-(2-trifluoromethyl-10-phenothiazinylpropyl]piperazine dihydrochloride, 10-[3-[4-(2-Hydroxyethyl)piperazinyl]propyl]-2-trifluoromethylphenothiazine dihydrochloride, 4-[3-[2-(Trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, Anatensol, Dapotum, Fludecan, Flufenazin, Fluorophenazine dihydrochloride, Fluphenazine chloride, Fluphenazine dihydrochloride, Fluphenazine hydrochloride, Liogen, Lyogen, Lyorodin, Mirenil, Moditen, OMCA, Permitil, Permitil hydrochloride, Prolinate, Prolixin, Siqualone, Squibb 4918, Tensofin, Trancin, Valamina
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Cl.OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1
IUPAC Name 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol;dihydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 510.44
Quantity 100 mg
Formula Weight 509.1282 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.