missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluphenazine.2HCl, Lipomed
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Fluphenazine.2HCl
Specifications
Specifications
| Chemical Name or Material | Fluphenazine dihydrochloride |
| CAS | 146-56-5 |
| Analyte or Component Names | Fluphenazine dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H26 F3 N3 O S . 2 Cl H |
| Synonym | Fluphenazine Dihydrochloride, 4-[3-[2-(Trifluoromethyl)-10 H-phenothiazin-10-yl]propyl]-1-piperazineethanol hydrochloride (1:2), 4-[3-[2-(Trifluoromethyl)-10 H-phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, 1-(2-Hydroxyethyl)-4-[3-(2-trifluoromethyl-10-phenothiazinylpropyl]piperazine dihydrochloride, 10-[3-[4-(2-Hydroxyethyl)piperazinyl]propyl]-2-trifluoromethylphenothiazine dihydrochloride, 4-[3-[2-(Trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, Anatensol, Dapotum, Fludecan, Flufenazin, Fluorophenazine dihydrochloride, Fluphenazine chloride, Fluphenazine dihydrochloride, Fluphenazine hydrochloride, Liogen, Lyogen, Lyorodin, Mirenil, Moditen, OMCA, Permitil, Permitil hydrochloride, Prolinate, Prolixin, Siqualone, Squibb 4918, Tensofin, Trancin, Valamina |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Cl.OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol;dihydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction