CAS RN 1431980-60-7
CAS RN 1431980-60-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Promotions Available
Tocris Bioscience™ TASP 0433864
Tocris Bioscience™ TASP 0433864
Selective mGlu2 receptor positive allosteric modulator
2
3