Learn More
Tocris Bioscience™ TASP 0433864, Tocris Bioscience™
GREENER_CHOICE
Selective mGlu2 receptor positive allosteric modulator
Brand:  Tocris Bioscience™ 5362/50
Encompass
Product Code. 15716756
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TASP 0433864 is a selective mGlu2 receptor positive allosteric modulator (EC50 values are 199 and 206 nM for human and rat receptors, respectively). Exhibits negligible activity against a range of neurotransmitter targets, except 5-HT2B and MAO-B (72 and 92% inhibition at 10 μM, respectively). Potentiates inhibitory effect of DCG IV (Cat. No. 0975) on excitatory postsynaptic potentials in hippocampal slices and ketamine- and methamphetamine-induced increases in locomotor activity in rats and mice.
Specifications
Specifications
(2S)-2-[[4-(1,1-Dimethylethyl)phenoxy]methyl]-2,3-dihydro-5-methylimidazo[2,1-b]oxazole-6-carboxamide
mGlu2 receptor positive allosteric modulator
Soluble to 100mM in DMSO and to 20mM in ethanol
White
1431980-60-7
50 mg
Solid
Bioactive Small Molecule
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
For Research Use Only.