CAS RN 143-33-9
Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N
Sodium cyanide, 98%, for analysis, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 SMILES: [Na+].[C-]#N
Sodium cyanide, 95+%, ACS reagent, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 SMILES: [Na+].[C-]#N
Sodium cyanide, granules, 98%, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N
Sodium Cyanide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Sodium Cyanide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Sodium Cyanide, ACS Reagent, ≥95.0%, Solstice
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N
Sodium Cyanide, Supelco™
CAS: 143-33-9
Sodium Cyanide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Sodium Cyanide, Reagent Grade, 97%, Honeywell™
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N