Product Code. 15681750
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
15681750
2.5 kg
Poly Bottle
2.5kg
15671750
100 g
Poly Bottle
100g
15691750
500 g
Poly Bottle
500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15681750
Supplier
Honeywell-Fluka
Supplier No.
2052222.5KG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
ACS Reagent,≥95.0%
Leading the Industry For Over 65 Years Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
CAS
143-33-9
Molecular Formula
CNNa
Molecular Weight (g/mol)
49.01
MDL Number
MFCD00003523
InChI Key
MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID
8929
IUPAC Name
sodium;cyanide
SMILES
[Na+].[C-]#N
Specifications
Boiling Point
1407°C
Melting Point
563°C
Color
Colorless
pH
11 to 12 (49g/L at 25°C)
Physical Form
Crystalline Powder
Quantity
2.5 kg
Linear Formula
NaCN
UN Number
UN1689
Beilstein
3587243
Solubility Information
Soluble in water
Formula Weight
49.01g/mol
Percent Purity
≥95.0%
Grade
ACS Reagent
Packaging
Poly Bottle
Appearance
Crystals
Vapor Pressure
1mmHg (817°C)
Density
1.6g/cu.cm; Vapor Density: 1.7 (vs air)
Chemical Name or Material
Sodium Cyanide
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?