missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyanide, ≥95%, (ACS Reagent Grade), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 143-33-9 |
| Quantity | 100 g |
| Assay Percent Range | ≥95% |
| Molecular Formula | CNNa |
| MDL Number | MFCD00003523 |
| Synonym | sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h |
| InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
| SMILES | [Na+].[C-]#N |
| IUPAC Name | sodium iminomethanide |
| Molecular Weight (g/mol) | 49.01 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction